Assay Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021305.3 | NP_067280.1 | 1 - 2 | 96 | 75 | |
| GenBank mRNA | AK017615.1 | - | 1 - 2 | 78 | 75 | |
| AK077701.1 | - | 1 - 2 | 96 | 75 | ||
| AK160602.1 | - | 1 - 2 | 82 | 75 | ||
| AK161316.1 | - | 1 - 2 | 175 | 75 | ||
| BC005458.1 | - | 1 - 2 | 39 | 3494001 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017615.1, AK077701.1, AK161316.1, BC005458.1, NM_021305.3 transcripts.
Set Membership: |
|

Sec61a2
Genomic Map