Assay Details
| Gene Symbol: Grin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177656.2 | NP_001171127.1 | 1 - 2 | 556 | 65 | |
| NM_001177657.2 | NP_001171128.1 | 1 - 2 | 556 | 65 | ||
| NM_008169.3 | NP_032195.1 | 1 - 2 | 556 | 65 | ||
| GenBank mRNA | AK036204.1 | - | 1 - 2 | 496 | 65 | |
| AK090286.1 | - | 1 - 2 | 536 | 65 | ||
| AK158140.1 | - | 1 - 2 | 496 | 65 | ||
| AK158175.1 | - | 1 - 2 | 494 | 65 | ||
| BC039157.1 | - | 1 - 2 | 472 | 4507986 | 65 | |
| CD355382.1 | - | 1 - 2 | 556 | 65 | ||
| D10028.1 | - | 1 - 2 | 348 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grin1
Genomic Map