Assay Details
Gene Symbol: Med19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025885.3 | NP_080161.1 | 3 - 4 | 614 | 145 | |
GenBank mRNA | AK005399.1 | - | 3 - 4 | 582 | 145 | |
AK010552.1 | - | 3 - 4 | 592 | 145 | ||
AK160579.1 | - | 3 - 4 | 580 | 145 | ||
AK168787.1 | - | 3 - 4 | 582 | 145 | ||
BB611514.1 | - | 3 - 4 | 614 | 145 | ||
BC025475.1 | - | 2 - 3 | 188 | 145 | ||
BC044207.1 | - | 3 - 4 | 476 | 145 | ||
BC044302.1 | - | 3 - 4 | 476 | 145 | ||
BC082559.1 | - | 3 - 4 | 579 | 30606373 | 145 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21393981 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21393980 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21393982 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | BC025475.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | AK139430.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | AW491437.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | NM_025885.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | AK010552.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | BC025475.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | AK139430.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | AW491437.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | NM_025885.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172842 | AK010552.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645441 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626181 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 973760 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208137 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833651 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49554 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | BC025475.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | AK139430.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | AW491437.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | NM_025885.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | AK010552.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | BC025475.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | AK139430.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | AW491437.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | NM_025885.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172842 | AK010552.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393985 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393984 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393987 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393992 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393990 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394002 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393982 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394005 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21394004 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393986 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393998 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393993 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21393988 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04591163 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04591157 | - | - |
Whole Mouse Genome G4122A | A_52_P453577 | BC082559.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P453577 | BC025475.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P453577 | BC044207.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P453577 | BC044302.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P453577 | AK139430.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | BC025475.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | AK139430.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | AW491437.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | NM_025885.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | AK010552.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | BC025475.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | AK139430.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | AW491437.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | NM_025885.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P172842 | AK010552.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137749 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124824 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01159284 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01279400 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01159283 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01279403 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21393982 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21393981 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21393980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393997 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393983 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21393993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21394010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21394010 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21394013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02162268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21394002 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393978 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01861071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01964284 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21394004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01478130 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393998 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393987 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01279412 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21394017 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393997 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21394005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21394009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21394008 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21393992 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10473347 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10473344 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10473346 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10473348 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10473345 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452395_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1427332_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10473343 | - | - |
Mouse Genome 430 2.0 Array | 1452395_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1427332_at | - | - |
Mouse Genome 430 2.0 Array | 1427332_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1452395_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381379_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 381379_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200009715.mm.1 | NM_025885.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009715.mm.1 | NM_025885.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009715.mm.1 | TC0200001218.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200009713.mm.1 | NM_025885.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009713.mm.1 | NM_025885.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009713.mm.1 | TC0200001218.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200009712.mm.1 | NM_025885.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009712.mm.1 | NM_025885.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009712.mm.1 | TC0200001218.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200009714.mm.1 | NM_025885.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009714.mm.1 | NM_025885.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200009714.mm.1 | TC0200001218.mm.1 | MTA TC |
Mouse Expression Set 430 | 1427332_at | - | - |
Mouse Expression Set 430 | 1427332_PM_at | - | - |
Mouse Expression Set 430 | 1452395_at | - | - |
Mouse Expression Set 430 | 1452395_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427332_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1427332_at | - | - |
Mouse Genome 430A 2.0 Array | 1452395_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452395_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452395_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452395_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427332_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1427332_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 102570014 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1214141 | - | - |
mouse-6 v1.1 (Illumina) | scl0381379.5_141-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102570014 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102570014 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2820098 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2820098 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2654541 | - | - |
