Assay Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175300.4 | NP_780509.2 | 11 - 12 | 2115 | 66 | |
| GenBank mRNA | AA727217.1 | - | 4 - 5 | 443 | 66 | |
| AK129351.1 | - | 11 - 12 | 2068 | 66 | ||
| AK154465.1 | - | 11 - 12 | 2069 | 66 | ||
| AK170676.1 | - | 11 - 12 | 2081 | 66 | ||
| BC027351.1 | - | 11 - 12 | 2063 | 66 | ||
| BC038267.1 | - | 10 - 11 | 1426 | 66 | ||
| BC065421.1 | - | 2 - 3 | 228 | 66 | ||
| BC096381.1 | - | 10 - 11 | 2308 | 66 | ||
| BC137583.1 | - | 11 - 12 | 2083 | 8860602 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc2
Genomic Map