Assay Details
| Gene Symbol: Anapc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175300.4 | NP_780509.2 | 5 - 6 | 1264 | 84 | |
| GenBank mRNA | AK033784.1 | - | 5 - 6 | 1232 | 84 | |
| AK129351.1 | - | 5 - 6 | 1217 | 84 | ||
| AK154465.1 | - | 5 - 6 | 1218 | 84 | ||
| AK170676.1 | - | 5 - 6 | 1230 | 84 | ||
| AK189707.1 | - | 3 - 4 | 345 | 84 | ||
| AK220035.1 | - | 1 - 2 | 100 | 84 | ||
| BC027351.1 | - | 5 - 6 | 1212 | 84 | ||
| BC038267.1 | - | 4 - 5 | 576 | 84 | ||
| BC096381.1 | - | 4 - 5 | 1457 | 84 | ||
| BC137583.1 | - | 5 - 6 | 1232 | 8860602 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Anapc2
Genomic Map