Assay Details
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009157.4 | NP_033183.1 | 6 - 7 | 737 | 128 | |
GenBank mRNA | AK036928.1 | - | 6 - 7 | 741 | 128 | |
AK044534.1 | - | 6 - 7 | 729 | 128 | ||
AK044698.1 | - | 6 - 7 | 729 | 128 | ||
AK139777.1 | - | 7 - 8 | 760 | 128 | ||
BC029833.1 | - | 4 - 5 | 488 | 3962163 | 128 | |
U18310.1 | - | 6 - 7 | 712 | 128 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
