Assay Details
| Gene Symbol: Hc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010406.2 | NP_034536.1 | 16 - 17 | 2125 | 135 | |
| GenBank mRNA | AK080166.1 | - | 16 - 17 | 2124 | 135 | |
| M35525.1 | - | 16 - 17 | 2080 | 135 | ||
| M35526.1 | - | 16 - 17 | 2078 | 135 | ||
More Information
Set Membership: |
|

Hc
Genomic Map