Assay Details
| Gene Symbol: Dab2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001602.2 | NP_001001602.2 | 8 - 9 | 1954 | 62 | |
| NM_001114124.2 | NP_001107596.1 | 10 - 11 | 2078 | 62 | ||
| NM_001114125.1 | NP_001107597.1 | 10 - 11 | 1930 | 62 | ||
| NM_001290637.1 | NP_001277566.1 | 10 - 11 | 1985 | 62 | ||
| NM_001290639.1 | NP_001277568.1 | 8 - 9 | 1613 | 62 | ||
| NM_001290640.1 | NP_001277569.1 | 8 - 9 | 1918 | 62 | ||
| NM_001290641.1 | NP_001277570.1 | 10 - 11 | 2211 | 62 | ||
| GenBank mRNA | AK122548.1 | - | 5 - 6 | 918 | 62 | |
| AK147464.1 | - | 10 - 11 | 2010 | 62 | ||
| AK147593.1 | - | 10 - 11 | 2212 | 62 | ||
| AK155296.1 | - | 10 - 11 | 1986 | 62 | ||
| AY305656.1 | - | 8 - 9 | 1671 | 62 | ||
| AY305657.1 | - | 8 - 9 | 1954 | 62 | ||
| AY305658.1 | - | 8 - 9 | 1613 | 62 | ||
| BC094373.1 | - | 5 - 6 | 775 | 62 | ||
| BC118530.1 | - | 8 - 9 | 1579 | 40098202 | 62 | |
| DQ473307.1 | - | 10 - 11 | 1815 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dab2ip
Genomic Map