Assay Details
Gene Symbol: Ndufa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026703.2 | NP_080979.1 | 2 - 3 | 265 | 68 | |
GenBank mRNA | AK002744.1 | - | 2 - 3 | 265 | 68 | |
AK088463.1 | - | 2 - 3 | 311 | 68 | ||
BC012416.1 | - | 2 - 3 | 263 | 3584066 | 68 | |
BC024673.1 | - | 2 - 3 | 262 | 4195454 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
