Assay Details
| Gene Symbol: Ptgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008969.4 | NP_032995.1 | 4 - 5 | 499 | 78 | |
| GenBank mRNA | AK046457.1 | - | 4 - 5 | 426 | 78 | |
| AK134163.1 | - | 4 - 5 | 385 | 78 | ||
| AK138432.1 | - | 4 - 5 | 438 | 78 | ||
| AK142741.1 | - | 4 - 5 | 426 | 78 | ||
| AK159907.1 | - | 4 - 5 | 500 | 78 | ||
| AK160886.1 | - | 4 - 5 | 418 | 78 | ||
| AK163609.1 | - | 4 - 5 | 426 | 78 | ||
| AK167357.1 | - | 3 - 4 | 377 | 78 | ||
| AK170414.1 | - | 4 - 5 | 394 | 78 | ||
| AK170664.1 | - | 4 - 5 | 426 | 78 | ||
| AY547265.1 | - | 3 - 4 | 467 | 78 | ||
| BC005573.1 | - | 4 - 5 | 416 | 3968101 | 78 | |
| BC023322.1 | - | 4 - 5 | 408 | 3968136 | 78 | |
| CT010314.1 | - | 3 - 4 | 362 | 78 | ||
| M34141.1 | - | 4 - 5 | 398 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK167357.1 transcripts.
Set Membership: |
|

Ptgs1
Genomic Map