Assay Details
| Gene Symbol: Ptgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008969.4 | NP_032995.1 | 8 - 9 | 1151 | 83 | |
| GenBank mRNA | AK046457.1 | - | 8 - 9 | 1078 | 83 | |
| AK134163.1 | - | 8 - 9 | 1037 | 83 | ||
| AK138432.1 | - | 8 - 9 | 1090 | 83 | ||
| AK142741.1 | - | 8 - 9 | 1078 | 83 | ||
| AK159907.1 | - | 8 - 9 | 1152 | 83 | ||
| AK160886.1 | - | 8 - 9 | 1070 | 83 | ||
| AK163609.1 | - | 8 - 9 | 1078 | 83 | ||
| AK167357.1 | - | 7 - 8 | 1029 | 83 | ||
| AK170414.1 | - | 8 - 9 | 1046 | 83 | ||
| AK170664.1 | - | 8 - 9 | 1078 | 83 | ||
| BC005573.1 | - | 8 - 9 | 1068 | 3968101 | 83 | |
| BC023322.1 | - | 8 - 9 | 1060 | 3968136 | 83 | |
| CT010314.1 | - | 7 - 8 | 1014 | 83 | ||
| M34141.1 | - | 8 - 9 | 1050 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptgs1
Genomic Map