Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200028687.mm.1 |
NM_145522.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028687.mm.1 |
NM_145522.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028687.mm.1 |
TC0200003350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200028696.mm.1 |
NM_145522.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028696.mm.1 |
NM_145522.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028696.mm.1 |
TC0200003350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200028690.mm.1 |
NM_145522.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028690.mm.1 |
NM_145522.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028690.mm.1 |
TC0200003350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200028695.mm.1 |
NM_145522.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028695.mm.1 |
NM_145522.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028695.mm.1 |
TC0200003350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200028697.mm.1 |
NM_145522.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028697.mm.1 |
NM_145522.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200028697.mm.1 |
TC0200003350.mm.1 |
MTA TC |