Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200000591.mm.1 |
NM_001198808.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000591.mm.1 |
NM_001198808.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000591.mm.1 |
NM_177386.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000591.mm.1 |
NM_177386.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000591.mm.1 |
TC0200000087.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200000592.mm.1 |
NM_001198808.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000592.mm.1 |
NM_001198808.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200000592.mm.1 |
TC0200000087.mm.1 |
MTA TC |