Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200024816.mm.1 |
NM_001165941.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024816.mm.1 |
NM_001165941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024816.mm.1 |
NM_001165943.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024816.mm.1 |
NM_001165943.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024816.mm.1 |
NM_028950.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024816.mm.1 |
NM_028950.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024816.mm.1 |
TC0200003005.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200024817.mm.1 |
NM_001165941.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024817.mm.1 |
NM_001165941.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024817.mm.1 |
NM_001165943.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024817.mm.1 |
NM_001165943.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024817.mm.1 |
NM_028950.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024817.mm.1 |
NM_028950.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200024817.mm.1 |
TC0200003005.mm.1 |
MTA TC |