Assay Details
| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153587.2 | NP_705815.1 | 4 - 5 | 651 | 112 | |
| GenBank mRNA | AK032316.1 | - | 4 - 5 | 744 | 112 | |
| AK046319.1 | - | 4 - 5 | 428 | 112 | ||
| AK135786.1 | - | 559 | 112 | |||
| AY341881.1 | - | 4 - 5 | 640 | 112 | ||
More Information
Set Membership: |
|

Rps6ka5
Genomic Map