Assay Details
| Gene Symbol: Trappc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024206.2 | NP_001019377.1 | 1 - 2 | 79 | 78 | |
| GenBank mRNA | CK621128.1 | - | 1 - 2 | 79 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001024206.2 transcripts.
Set Membership: |
|

Trappc1
Genomic Map