Assay Details
| Gene Symbol: Syt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016908.2 | NP_058604.1 | 5 - 6 | 965 | 117 | |
| GenBank mRNA | AB026806.1 | - | 5 - 6 | 709 | 117 | |
| BC047148.1 | - | 5 - 6 | 965 | 6465481 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syt5
Genomic Map