Assay Details
| Gene Symbol: Syt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016908.2 | NP_058604.1 | 7 - 8 | 1214 | 74 | |
| GenBank mRNA | AB026806.1 | - | 7 - 8 | 958 | 74 | |
| BC047148.1 | - | 7 - 8 | 1214 | 6465481 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
synaptic vesicle exocytosis
calcium ion regulated exocytosis
regulation of calcium ion-dependent exocytosis
synaptic vesicle endocytosis
calcium ion-regulated exocytosis of neurotransmitter
calcium ion binding
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding
metal ion binding
protein heterodimerization activity
synaptic vesicle exocytosis
calcium ion regulated exocytosis
regulation of calcium ion-dependent exocytosis
synaptic vesicle endocytosis
calcium ion-regulated exocytosis of neurotransmitter
calcium ion binding
calcium-dependent phospholipid binding
syntaxin binding
clathrin binding
metal ion binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117425 | BC047148.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117425 | NM_016908.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 95479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8078 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546689 | AK030736.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546689 | AK079866.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546689 | BC048179.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546689 | NM_145956.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151276 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 104153 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P177419 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117425 | BC047148.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117425 | NM_016908.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949395 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949381 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949397 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949389 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949415 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949412 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949391 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949405 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24949413 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P177419 | - | - |
| Whole Mouse Genome G4122A | A_52_P667324 | BC047148.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P667324 | NM_016908.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P667324 | AB026806.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129396 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P177419 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260413 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260416 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260421 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260415 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260418 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06260422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06122485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949351 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06260416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06002348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949385 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949373 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06260425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06654331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05979726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24949363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06260421 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559581 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559589 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559587 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559585 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559582 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559586 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559584 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559588 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10559583 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422531_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559580 | - | - |
| Mouse Genome 430 2.0 Array | 1422531_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422531_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53420_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 53420_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700019718.mm.1 | TC0700002162.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700019718.mm.1 | NM_016908.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700019718.mm.1 | NM_016908.2 | Refseq |
| Mouse Expression Set 430 | 1422531_at | - | - |
| Mouse Expression Set 430 | 1422531_PM_at | - | - |
| Murine Genome U74v2 Set | 104269_PM_at | - | - |
| Murine Genome U74v2 Set | 104269_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422531_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422531_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422531_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422531_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 307199 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4150082 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4150082 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004150082 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1220815 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220815 | - | - |
| mouse-6 v1.1 (Illumina) | scl31809.10.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4150082 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31809.10.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4150082 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1220815 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1220815 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl31809.10.1_30-S | - | - |

Syt5
Genomic Map