Assay Details
| Gene Symbol: Ppp1r12c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029834.3 | NP_084110.2 | 4 - 5 | 829 | 65 | |
| GenBank mRNA | AK046570.1 | - | 4 - 5 | 799 | 65 | |
| AK144691.1 | - | 4 - 5 | 830 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r12c
Genomic Map