Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100006393.mm.1 |
NM_001039536.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006393.mm.1 |
NM_001039536.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006393.mm.1 |
TC1100000798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006394.mm.1 |
NM_001039536.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006394.mm.1 |
NM_001039536.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006394.mm.1 |
TC1100000798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006391.mm.1 |
NM_001039536.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006391.mm.1 |
NM_001039536.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006391.mm.1 |
TC1100000798.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100006392.mm.1 |
NM_001039536.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006392.mm.1 |
NM_001039536.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100006392.mm.1 |
TC1100000798.mm.1 |
MTA TC |