Assay Details
Gene Symbol: Zim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011769.4 | NP_035899.4 | 1 - 2 | 60 | 86 | |
GenBank mRNA | AK086590.1 | - | 1 - 2 | 79 | 86 | |
BC066196.1 | - | 1 - 2 | 60 | 30430385 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK086590.1, BC066196.1, NM_011769.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
