Assay Details
| Gene Symbol: Tgfbi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009369.4 | NP_033395.1 | 14 - 15 | 1952 | 65 | |
| GenBank mRNA | AK084431.1 | - | 14 - 15 | 1970 | 65 | |
| AK142323.1 | - | 14 - 15 | 1972 | 65 | ||
| AK150449.1 | - | 14 - 15 | 1970 | 65 | ||
| AK150833.1 | - | 14 - 15 | 1970 | 65 | ||
| AK150969.1 | - | 14 - 15 | 1971 | 65 | ||
| AK151680.1 | - | 14 - 15 | 1967 | 65 | ||
| AK152365.1 | - | 14 - 15 | 1971 | 65 | ||
| AK152662.1 | - | 14 - 15 | 1970 | 65 | ||
| AK155572.1 | - | 14 - 15 | 1967 | 65 | ||
| AK155828.1 | - | 14 - 15 | 1967 | 65 | ||
| AK159711.1 | - | 14 - 15 | 1966 | 65 | ||
| AK164373.1 | - | 14 - 15 | 1970 | 65 | ||
| AK170495.1 | - | 14 - 15 | 1967 | 65 | ||
| AK171557.1 | - | 13 - 14 | 1791 | 65 | ||
| BC129900.1 | - | 14 - 15 | 1945 | 40111549 | 65 | |
| L19932.1 | - | 14 - 15 | 1953 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tgfbi
Genomic Map