Assay Details
Gene Symbol: Tfap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301674.1 | NP_001288603.1 | 2 - 3 | 393 | 93 | |
NM_011547.4 | NP_035677.2 | 1 - 2 | 149 | 93 | ||
GenBank mRNA | AK145772.1 | - | 1 - 2 | 104 | 93 | |
BC007471.1 | - | 1 - 2 | 124 | 3497082 | 93 | |
BC018226.1 | - | 2 - 3 | 392 | 3983850 | 93 | |
BY731472.1 | - | 1 - 2 | 301 | 93 | ||
U17285.1 | - | 1 - 2 | 141 | 93 | ||
X74216.1 | - | 1 - 2 | 110 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK145772.1, BC007471.1, BC018226.1, NM_001301674.1, NM_011547.4, U17285.1, X74216.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
