Assay Details
Gene Symbol: Pak6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033254.3 | NP_001028426.2 | 6 - 7 | 2003 | 77 | |
NM_001145854.1 | NP_001139326.1 | 5 - 6 | 1528 | 77 | ||
GenBank mRNA | AK028788.1 | - | 5 - 6 | 1512 | 77 | |
AK145601.1 | - | 5 - 6 | 1502 | 77 | ||
AK158913.1 | - | 6 - 7 | 2277 | 77 | ||
AK158944.1 | - | 6 - 7 | 2277 | 77 | ||
BC059912.1 | - | 6 - 7 | 1809 | 77 | ||
BC079572.1 | - | 6 - 7 | 2000 | 77 | ||
BC150754.1 | - | 6 - 7 | 1893 | 9087665 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
