Assay Details
| Gene Symbol: Bub1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009773.3 | NP_033903.2 | 6 - 7 | 972 | 104 | |
| GenBank mRNA | AF107296.1 | - | 6 - 7 | 927 | 104 | |
| AK133909.1 | - | 6 - 7 | 913 | 104 | ||
| BC031577.1 | - | 6 - 7 | 923 | 4953223 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bub1b
Genomic Map