Assay Details
| Gene Symbol: Ppp1r14d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290796.1 | NP_001277725.1 | 3 - 4 | 479 | 72 | |
| NM_028104.4 | NP_082380.1 | 2 - 3 | 414 | 72 | ||
| GenBank mRNA | AF408400.2 | - | 2 - 3 | 413 | 72 | |
| AK008348.1 | - | 2 - 3 | 372 | 72 | ||
| AY179331.1 | - | 3 - 4 | 466 | 72 | ||
| BC024547.1 | - | 2 - 3 | 413 | 4989982 | 72 | |
More Information
Set Membership: |
|

Ppp1r14d
Genomic Map