Assay Details
Gene Symbol: Cdyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009881.3 | NP_034011.1 | 3 - 4 | 362 | 72 | |
GenBank mRNA | AF081260.1 | - | 3 - 4 | 344 | 72 | |
AF081261.1 | - | 3 - 4 | 344 | 72 | ||
BC055103.1 | - | 3 - 4 | 311 | 6741468 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
