Assay Details
Gene Symbol: Cdyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123386.1 | NP_001116858.1 | 3 - 4 | 1411 | 131 | |
NM_009881.3 | NP_034011.1 | 6 - 7 | 1361 | 131 | ||
GenBank mRNA | AF081260.1 | - | 6 - 7 | 1343 | 131 | |
AF081261.1 | - | 6 - 7 | 1343 | 131 | ||
AK028162.1 | - | 3 - 4 | 1107 | 131 | ||
AK156509.1 | - | 3 - 4 | 1412 | 131 | ||
AK162180.1 | - | 1 - 2 | 91 | 131 | ||
BC055103.1 | - | 6 - 7 | 1310 | 6741468 | 131 | |
BC062123.1 | - | 3 - 4 | 1500 | 6826210 | 131 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
