Assay Details
| Gene Symbol: Olfr1044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147011.1 | NP_667222.1 | 1 | 464 | 83 | |
| GenBank mRNA | BC111826.1 | - | 1 | 463 | 83 | |
More Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			Set Membership: | 
		   
					
						
							 | 
				

Olfr1044
Genomic Map