Assay Details
| Gene Symbol: Lair1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113474.1 | NP_001106945.1 | 3 - 4 | 499 | 124 | |
| GenBank mRNA | AF479685.1 | - | 3 - 4 | 434 | 124 | |
| AF491829.1 | - | 3 - 4 | 532 | 124 | ||
| AF491831.1 | - | 3 - 4 | 411 | 124 | ||
| AK044465.1 | - | 3 - 4 | 470 | 124 | ||
| AK050281.1 | - | 3 - 4 | 455 | 124 | ||
| AK054158.1 | - | 3 - 4 | 578 | 124 | ||
| AY392763.1 | - | 3 - 4 | 436 | 124 | ||
| BC025901.1 | - | 3 - 4 | 463 | 5253846 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lair1
Genomic Map