Assay Details
| Gene Symbol: Lrrc4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289742.1 | NP_001276671.1 | 5 - 6 | 1042 | 66 | |
| GenBank mRNA | AK048322.1 | - | 5 - 6 | 996 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK048322.1, NM_001289742.1 transcripts.
Set Membership: |
|

Lrrc4c
Genomic Map