Assay Details
Gene Symbol: Lrrc4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289742.1 | NP_001276671.1 | 7 - 8 | 1188 | 149 | |
NM_001289743.1 | NP_001276672.1 | 6 - 7 | 972 | 149 | ||
GenBank mRNA | AK034276.1 | - | 6 - 7 | 973 | 149 | |
AK048322.1 | - | 7 - 8 | 1142 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK034276.1, AK048322.1, NM_001289742.1, NM_001289743.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
