Assay Details
| Gene Symbol: Olfr1349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207136.1 | NP_997019.1 | 1 | 619 | 83 | |
| GenBank mRNA | BC147396.1 | - | 1 | 641 | 9056466 | 83 |
| BC147397.1 | - | 1 | 641 | 9056451 | 83 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118259 | BC147396.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118259 | BC147397.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2118259 | NM_207136.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118259 | BC147396.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118259 | BC147397.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2118259 | NM_207136.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P317608 | BC147396.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P317608 | BC147397.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P317608 | NM_207136.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955879 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955887 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955837 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955881 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955875 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955890 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955841 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955848 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955858 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24955854 | - | - |
| Whole Mouse Genome G4122A | A_52_P317608 | BC147396.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P317608 | BC147397.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P317608 | NM_207136.1 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06206365 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06206369 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06206373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24955848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06611700 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06206355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955873 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955881 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06206360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06206359 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06261847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06206358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06413280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06261846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955891 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06290781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06206370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955839 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06366402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955871 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06121485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955859 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05990261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06121487 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955879 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955882 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955858 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06118190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955841 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955843 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24955856 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10559789 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10559788 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 603235 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1252154 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2888754 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2888754 | - | - |

Olfr1349
Genomic Map