Assay Details
| Gene Symbol: Olfr342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146948.1 | NP_667159.1 | 1 | 447 | 99 | |
| GenBank mRNA | BC120521.1 | - | 1 | 473 | 40129444 | 99 |
| BC120523.1 | - | 1 | 473 | 40129446 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Olfr342
Genomic Map