Assay Details
Gene Symbol: Lrch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081173.1 | NP_001074642.1 | 5 - 6 | 922 | 177 | |
GenBank mRNA | AK144916.1 | - | 5 - 6 | 752 | 177 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
