Assay Details
| Gene Symbol: Ppp1r13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011625.1 | NP_035755.1 | 3 - 4 | 465 | 76 | |
| GenBank mRNA | AK148849.1 | - | 494 | 76 | ||
| BC053092.1 | - | 3 - 4 | 491 | 5719383 | 76 | |
| BC054788.1 | - | 3 - 4 | 465 | 6826065 | 76 | |
More Information
Set Membership: |
|

Ppp1r13b
Genomic Map