Assay Details
| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291005.1 | NP_001277934.1 | 1 - 2 | 157 | 84 | |
| NM_019789.4 | NP_062763.2 | 1 - 2 | 157 | 84 | ||
| GenBank mRNA | AF287732.1 | - | 1 - 2 | 135 | 84 | |
| AF287733.1 | - | 1 - 2 | 135 | 84 | ||
| AF300870.1 | - | 1 - 2 | 80 | 84 | ||
| AK165711.1 | - | 1 - 2 | 105 | 84 | ||
| BC026980.1 | - | 1 - 2 | 94 | 5368268 | 84 | |
| BC047139.1 | - | 1 - 2 | 94 | 6389719 | 84 | |
| BC057329.1 | - | 1 - 2 | 76 | 6813819 | 84 | |
| CJ148267.1 | - | 1 - 2 | 84 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF287732.1, AF287733.1, AF300870.1, AK165711.1, BC026980.1, BC047139.1, BC057329.1, NM_019789.4 transcripts.
Set Membership: |
|

Kcnip3
Genomic Map