Assay Details
| Gene Symbol: Kcnip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111331.1 | NP_001104801.1 | 4 - 5 | 760 | 98 | |
| NM_001291005.1 | NP_001277934.1 | 5 - 6 | 582 | 98 | ||
| NM_019789.4 | NP_062763.2 | 5 - 6 | 586 | 98 | ||
| NR_110989.1 | 5 - 6 | 746 | 98 | |||
| GenBank mRNA | AF184624.1 | - | 5 - 6 | 534 | 98 | |
| AF274050.1 | - | 4 - 5 | 446 | 98 | ||
| AF287732.1 | - | 5 - 6 | 560 | 98 | ||
| AF287733.1 | - | 5 - 6 | 564 | 98 | ||
| AF300870.1 | - | 5 - 6 | 509 | 98 | ||
| AK016721.1 | - | 5 - 6 | 748 | 98 | ||
| AK165711.1 | - | 5 - 6 | 534 | 98 | ||
| BC026980.1 | - | 5 - 6 | 523 | 5368268 | 98 | |
| BC047139.1 | - | 5 - 6 | 523 | 6389719 | 98 | |
| BC057329.1 | - | 5 - 6 | 505 | 6813819 | 98 | |
| DQ148499.1 | - | 4 - 5 | 446 | 98 | ||
| DQ148500.1 | - | 4 - 5 | 760 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnip3
Genomic Map