Assay Details
Gene Symbol: Ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024209.2 | NP_077171.1 | 4 - 5 | 474 | 117 | |
GenBank mRNA | AK002764.1 | - | 4 - 5 | 436 | 117 | |
AK009104.1 | - | 4 - 5 | 454 | 117 | ||
BC002223.1 | - | 4 - 5 | 499 | 3491111 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
