Assay Details
| Gene Symbol: Slc22a23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033167.3 | NP_001028339.1 | 2 - 3 | 1132 | 80 | |
| GenBank mRNA | AK147397.1 | - | 2 - 3 | 1133 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a23
Genomic Map