Assay Details
| Gene Symbol: Agps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172666.3 | NP_766254.2 | 14 - 15 | 1550 | 99 | |
| GenBank mRNA | AK031049.1 | - | 14 - 15 | 1552 | 99 | |
| AK031154.1 | - | 14 - 15 | 1391 | 99 | ||
| AK079447.1 | - | 13 - 14 | 1230 | 99 | ||
| BC063086.1 | - | 14 - 15 | 1535 | 6332574 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Agps
Genomic Map