Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200006678.mm.1 |
NM_027990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006678.mm.1 |
NM_027990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006678.mm.1 |
TC0200000803.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006675.mm.1 |
NM_027990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006675.mm.1 |
NM_027990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006675.mm.1 |
TC0200000803.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006673.mm.1 |
NM_027990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006673.mm.1 |
NM_027990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006673.mm.1 |
TC0200000803.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006676.mm.1 |
NM_027990.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006676.mm.1 |
NM_027990.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006676.mm.1 |
TC0200000803.mm.1 |
MTA TC |