Assay Details
| Gene Symbol: Il2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008367.3 | NP_032393.3 | 2 - 3 | 455 | 83 | |
| GenBank mRNA | AK037369.1 | - | 2 - 3 | 441 | 83 | |
| AK088594.1 | - | 2 - 3 | 397 | 83 | ||
| AK171665.1 | - | 2 - 3 | 392 | 83 | ||
| BC114437.1 | - | 2 - 3 | 361 | 40054387 | 83 | |
| K02891.1 | - | 2 - 3 | 338 | 83 | ||
| M30856.1 | - | 2 - 3 | 385 | 83 | ||
| M54934.1 | - | 2 - 3 | 258 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Il2ra
Genomic Map