Assay Details
| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009804.2 | NP_033934.2 | 10 - 11 | 1436 | 76 | |
| GenBank mRNA | AK075853.1 | - | 10 - 11 | 1406 | 76 | |
| AK088168.1 | - | 10 - 11 | 1398 | 76 | ||
| AK128919.1 | - | 10 - 11 | 1411 | 76 | ||
| AK133881.1 | - | 10 - 11 | 1404 | 76 | ||
| AK148961.1 | - | 10 - 11 | 1404 | 76 | ||
| AK150893.1 | - | 10 - 11 | 1407 | 76 | ||
| AK159152.1 | - | 10 - 11 | 1407 | 76 | ||
| AK159885.1 | - | 10 - 11 | 1407 | 76 | ||
| AK159891.1 | - | 10 - 11 | 1358 | 76 | ||
| AK159913.1 | - | 10 - 11 | 1388 | 76 | ||
| AK169069.1 | - | 10 - 11 | 1401 | 76 | ||
| BC013447.1 | - | 10 - 11 | 1393 | 4241333 | 76 | |
| BC047126.1 | - | 10 - 11 | 1351 | 76 | ||
More Information
Set Membership: |
|

Cat
Genomic Map