Assay Details
| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009804.2 | NP_033934.2 | 4 - 5 | 587 | 82 | |
| GenBank mRNA | AK075853.1 | - | 4 - 5 | 557 | 82 | |
| AK088168.1 | - | 4 - 5 | 549 | 82 | ||
| AK128919.1 | - | 4 - 5 | 562 | 82 | ||
| AK133881.1 | - | 4 - 5 | 555 | 82 | ||
| AK148961.1 | - | 4 - 5 | 555 | 82 | ||
| AK150893.1 | - | 4 - 5 | 558 | 82 | ||
| AK159885.1 | - | 4 - 5 | 558 | 82 | ||
| AK159891.1 | - | 4 - 5 | 509 | 82 | ||
| AK159913.1 | - | 4 - 5 | 539 | 82 | ||
| AK169069.1 | - | 4 - 5 | 552 | 82 | ||
| AK188260.1 | - | 3 - 4 | 271 | 82 | ||
| AK188401.1 | - | 3 - 4 | 271 | 82 | ||
| AY040626.1 | - | 4 - 5 | 528 | 82 | ||
| BC013447.1 | - | 4 - 5 | 544 | 4241333 | 82 | |
| BC047126.1 | - | 4 - 5 | 502 | 82 | ||
| L25069.1 | - | 4 - 5 | 559 | 82 | ||
| M62897.1 | - | 4 - 5 | 472 | 82 | ||
| X52108.1 | - | 4 - 5 | 472 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cat
Genomic Map