Assay Details
| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009804.2 | NP_033934.2 | 5 - 6 | 697 | 60 | |
| GenBank mRNA | AK075853.1 | - | 5 - 6 | 667 | 60 | |
| AK088168.1 | - | 5 - 6 | 659 | 60 | ||
| AK128919.1 | - | 5 - 6 | 672 | 60 | ||
| AK133881.1 | - | 5 - 6 | 665 | 60 | ||
| AK148961.1 | - | 5 - 6 | 665 | 60 | ||
| AK150893.1 | - | 5 - 6 | 668 | 60 | ||
| AK159152.1 | - | 5 - 6 | 668 | 60 | ||
| AK159885.1 | - | 5 - 6 | 668 | 60 | ||
| AK159891.1 | - | 5 - 6 | 619 | 60 | ||
| AK159913.1 | - | 5 - 6 | 649 | 60 | ||
| AK169069.1 | - | 5 - 6 | 662 | 60 | ||
| AK216835.1 | - | 1 - 2 | 37 | 60 | ||
| AY040626.1 | - | 5 - 6 | 638 | 60 | ||
| BC013447.1 | - | 5 - 6 | 654 | 4241333 | 60 | |
| BC047126.1 | - | 5 - 6 | 612 | 60 | ||
| L25069.1 | - | 5 - 6 | 669 | 60 | ||
| M62897.1 | - | 5 - 6 | 582 | 60 | ||
| X52108.1 | - | 5 - 6 | 582 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cat
Genomic Map