Assay Details
Gene Symbol: Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009804.2 | NP_033934.2 | 9 - 10 | 1307 | 67 | |
GenBank mRNA | AK075853.1 | - | 9 - 10 | 1277 | 67 | |
AK088168.1 | - | 9 - 10 | 1269 | 67 | ||
AK128919.1 | - | 9 - 10 | 1282 | 67 | ||
AK133881.1 | - | 9 - 10 | 1275 | 67 | ||
AK148961.1 | - | 9 - 10 | 1275 | 67 | ||
AK150893.1 | - | 9 - 10 | 1278 | 67 | ||
AK159152.1 | - | 9 - 10 | 1278 | 67 | ||
AK159885.1 | - | 9 - 10 | 1278 | 67 | ||
AK159891.1 | - | 9 - 10 | 1229 | 67 | ||
AK159913.1 | - | 9 - 10 | 1259 | 67 | ||
AK169069.1 | - | 9 - 10 | 1272 | 67 | ||
AY040626.1 | - | 9 - 10 | 1248 | 67 | ||
BC013447.1 | - | 9 - 10 | 1264 | 4241333 | 67 | |
BC047126.1 | - | 9 - 10 | 1222 | 67 | ||
L25069.1 | - | 9 - 10 | 1279 | 67 | ||
M62897.1 | - | 9 - 10 | 1192 | 67 | ||
X52108.1 | - | 9 - 10 | 1192 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
