Assay Details
| Gene Symbol: Ubqln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018798.2 | NP_061268.2 | 1 | 1677 | 115 | |
| GenBank mRNA | AF177346.1 | - | 1 | 1678 | 115 | |
| AK076159.1 | - | 1 | 1681 | 115 | ||
| BC021824.1 | - | 1 | 1059 | 115 | ||
| BC053022.1 | - | 1 | 1677 | 5708544 | 115 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ubqln2
Genomic Map