Assay Details
| Gene Symbol: Nr2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010150.2 | NP_034280.2 | 2 - 3 | 1058 | 133 | |
| GenBank mRNA | AK131941.1 | - | 2 - 3 | 1022 | 133 | |
| AK146779.1 | - | 2 - 3 | 762 | 133 | ||
| AK171552.1 | - | 2 - 3 | 555 | 133 | ||
| BC008138.1 | - | 2 - 3 | 777 | 3582557 | 133 | |
| X76654.1 | - | 2 - 3 | 577 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr2f6
Genomic Map