Assay Details
| Gene Symbol: Dda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294258.1 | NP_001281187.1 | 1 - 2 | 137 | 55 | |
| NM_025600.2 | NP_079876.1 | 1 - 2 | 137 | 55 | ||
| NR_122114.1 | 1 - 2 | 137 | 55 | |||
| GenBank mRNA | AA036394.1 | - | 1 - 2 | 113 | 55 | |
| AK006318.1 | - | 1 - 2 | 121 | 55 | ||
| AK032966.1 | - | 1 - 2 | 95 | 55 | ||
| AK149117.1 | - | 1 - 2 | 119 | 55 | ||
| AK160132.1 | - | 1 - 2 | 133 | 55 | ||
| AK168085.1 | - | 1 - 2 | 95 | 55 | ||
| BB610194.1 | - | 1 - 2 | 130 | 55 | ||
| BC034701.1 | - | 1 - 2 | 97 | 3494430 | 55 | |
| BC093523.1 | - | 1 - 2 | 80 | 3711509 | 55 | |
| BY316490.1 | - | 1 - 2 | 55 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006318.1, AK032966.1, AK168085.1, BC034701.1, BC093523.1, NM_001294258.1, NM_025600.2 transcripts.
Set Membership: |
|

Dda1
Genomic Map